Learn More
Aviva Systems Biology Corp TMEM187 Antibody - middle region (ARP45476_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45476_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM187. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM187. Alternate names: CXorf12, DXS9878E, ITBA1ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-59035