Learn More
Aviva Systems Biology Corp TMEM195 Antibody - N-terminal region (ARP44468_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44468_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM195. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM195. Alternate names: FLJ16237, MGC131748, TMEM195ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58820