Learn More
Aviva Systems Biology Corp TMEM195 Antibody - middle region (ARP44470_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44470_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM195. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM195. Alternate names: FLJ16237, MGC131748, TMEM195ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62833