Learn More
Aviva Systems Biology Corp CEMIP2 Antibody - middle region (ARP42382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMEM2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58206