Learn More
Aviva Systems Biology Corp TMEM24 Antibody - C-terminal region (ARP47081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM24. Alternate names: C2CD2L, DLNB23, KIAA0285, TMEM24ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53502