Learn More
Aviva Systems Biology Corp TMEM30A Antibody - middle region (ARP47410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM30A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human TMEM30A. Alternate names: C6orf67, CDC50A, FLJ10856ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53514