Learn More
Aviva Systems Biology Corp Tmem33 Antibody - middle region (ARP47398_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47398_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tmem33. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Tmem33ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-301