Learn More
Aviva Systems Biology Corp TMEM35A Antibody - C-terminal region (ARP49658_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM35. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM35. Alternate names: FLJ14084ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60139