Learn More
Aviva Systems Biology Corp TMEM38A Antibody - N-terminal region (ARP49799_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49799_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM38A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM38A. Alternate names: MGC3169, TRICA, TRIC-AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-60188