Learn More
Aviva Systems Biology Corp TMEM42 Antibody - N-terminal region (ARP52709_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52709_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM42. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM42. Alternate names: MGC29956ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rabbit
Catalog No. 50-014-62908