Learn More
ABclonal Technology TMEM43 Rabbit mAb
SDP
Catalog No. 5000007853
Encompass_Preferred
This gene belongs to the TMEM43 family Defects in this gene are the cause of familial arrhythmogenic right ventricular dysplasia type 5 (ARVD5) also known as arrhythmogenic right ventricular cardiomyopathy type 5 (ARVC5) Arrhythmogenic right ventricular dysplasia is an inherited disorder often involving both ventricles and is characterized by ventricular tachycardia heart failure sudden cardiac death and fibrofatty replacement of cardiomyocytes This gene contains a response element for PPAR gamma (an adipogenic transcription factor) which may explain the fibrofatty replacement of the myocardium a characteristic pathological finding in ARVC
Catalog No. 50-000-07853 Supplier ABclonal Technology Supplier No. A21160100UL
This gene belongs to the TMEM43 family Defects in this gene are the cause of familial arrhythmogenic right ventricular dysplasia type 5 (ARVD5) also known as arrhythmogenic right ventricular cardiomyopathy type 5 (ARVC5) Arrhythmogenic right ventricular dysplasia is an inherited disorder often involving both ventricles and is characterized by ventricular tachycardia heart failure sudden cardiac death and fibrofatty replacement of cardiomyocytes This gene contains a response element for PPAR gamma (an adipogenic transcription factor) which may explain the fibrofatty replacement of the myocardium a characteristic pathological finding in ARVC