Learn More
Aviva Systems Biology Corp Tmem50b Antibody - N-terminal region (ARP60093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tmem50b. It was validated on Western Blot. . Alternate names: AU015466, AU019872, B230114J08RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-937