Learn More
Aviva Systems Biology Corp TMEM51 Antibody - N-terminal region (ARP47388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM51. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM51. Alternate names: C1orf72, FLJ10199ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-59535