Learn More
Aviva Systems Biology Corp MACO1 Antibody - N-terminal region (ARP47404_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM57. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM57. Alternate names: FLJ10747, FLJ23007ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53513