Learn More
Aviva Systems Biology Corp TMEM63A Antibody - C-terminal region (ARP40785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM63A. It was validated on Western Blot. . Alternate names: KIAA0489, KIAA0792ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-891