Learn More
Aviva Systems Biology Corp TMEM63A Antibody - N-terminal region (ARP40784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM63A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM63A. Alternate names: KIAA0489, KIAA0792, RP4-559A3.1ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-53236