Learn More
Aviva Systems Biology Corp TMEM69 Antibody - middle region (ARP47258_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47258_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM69. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TMEM69. Alternate names: C1orf154, FLJ21029, MGC104183, RP11-767N6.4ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53510