Learn More
Aviva Systems Biology Corp TMEM75 Antibody - C-terminal region (ARP44839_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM75. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TMEM75. Alternate names: FLJ36105ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58914