Learn More
Aviva Systems Biology Corp TMEM91 Antibody - N-terminal region (ARP44955_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMEM91. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMEM91. Alternate names: FLJ27310, FLJ45695, IFITMD6ApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rat
Catalog No. 50-014-58945