Learn More
Aviva Systems Biology Corp TMF1 Antibody - middle region (P100705_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100705_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMF1. Alternate names: ARA160, TMFApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62688