Learn More
MSC TMI, LLC Replacement Dock Curtain Roll, 8 in x 150 ft, 0.08 in, Clear PVC
Catalog No. 5000531255
Encompass
This replacement dock curtain roll provides a transparent, smooth barrier, constructed from clear polyvinylchloride (PVC). It measures 8 inches wide, 150 feet long, and has a thickness of 0.08 inches, making it suitable for a variety of standard duty industrial applications.
  • Features a smooth style.
  • Clear transparency for visibility.
  • Constructed from durable PVC material.
  • Designed for standard duty applications.
  • Standard mount type.
  • Not suitable for low temperature environments.
  • Not antistatic or reinforced.
  • Not intended for outdoor use.
Catalog No. 50-005-31255 Supplier MSC Supplier No. 57336901
This replacement dock curtain roll provides a transparent, smooth barrier, constructed from clear polyvinylchloride (PVC). It measures 8 inches wide, 150 feet long, and has a thickness of 0.08 inches, making it suitable for a variety of standard duty industrial applications.
  • Features a smooth style.
  • Clear transparency for visibility.
  • Constructed from durable PVC material.
  • Designed for standard duty applications.
  • Standard mount type.
  • Not suitable for low temperature environments.
  • Not antistatic or reinforced.
  • Not intended for outdoor use.