Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMIGD2 Detection Set (Risk Free), ProSci
SDP

Catalog No. 16595211 Shop All ProSci Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-211 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-211 Supplier ProSci LLC Supplier No. RF16120SET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Detection Sets

TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for ELISA, Western Blot, ICC, IHC-P, Flow, IF

TMIGD2 Antibody: TMIGD2 (transmembrane and immunoglobulin domain containing 1), also known as immunoglobulin-containing and proline-rich receptor 1 (IGPR1), is novel adhesion molecule that is expressed in multiple tissues, primarily in cells of epithelium and endothelium origins (1). TMIGD2 is thought to be involved in angiogenesis and regulates cellular morphology, homophilic cell aggregation, and cell-cell interaction. TMIGD2 activity also modulates actin stress fiber formation and focal adhesion and reduces cell migration. ProSci's Risk-FreeTM antibodies are mouse monoclonal antibodies made to improve in vivo studies. Unlike antibodies developed using proteins made in yeast or bacteria, Risk-FreeTM antibodies are developed with antigens expressed in mammalian cell lines, giving the most native post-translational modifications to the protein. Validated for flow cytometry and ELISA Rigorously tested for the following applications: Immunoblot Immunohistochemistry Immunocytochemistry Immunofluorescence Multiple antibodies per target allowing the user to choose the best antibody for their application Available individually or as a set Risk-FreeTM means they are guaranteed to work for their approved applications

Specifications

Antigen TMIGD2
Applications ELISA, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
Classification Monoclonal
Concentration Antibody 1 mg/mL
Conjugate Unconjugated
Formulation PBS containing 0.02% sodium azide.
Host Species Mouse
Purification Method Antibodies are supplied as protein A purified IgG1.
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Antibodies can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Detection Sets
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.