Learn More
Enzo Life Sciences TMIGD2 monoclonal antibody (8A1) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7030100
Encompass_Preferred
Clone: 8A1. Isotype: IgG1κ. Immunogen: Extracellular domain of human TMIGD2. Species reactivity: Human. Applications: Flow Cytometry, ICC, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide and 50% glycerol.
Catalog No. 50-201-3242