Learn More
Medchemexpress LLC TML-6 | 1462868-88-7 | 99.0% | C30H37NO7 | 10MG
SDP
Catalog No. 5000261215
Encompass_Preferred
TML-6 is an orally active curcumin-derived research compound that inhibits synthesis of the β-amyloid precursor protein and β-amyloid (Aβ). It modulates ApoE, suppresses NF-κB and mTOR signaling, and enhances Nrf2-mediated antioxidant activity, supporting its use in Alzheimer's disease and neurodegeneration research.
  • Orally active curcumin derivative
  • Inhibits β-amyloid precursor protein and Aβ synthesis
  • Modulates ApoE expression
  • Suppresses NF-κB and mTOR signaling pathways
  • Enhances Nrf2-mediated antioxidant activity
  • Available as solid and DMSO solution pack sizes for convenience
Catalog No. 50-002-61215 Supplier Medchemexpress LLC Supplier No. HY13731510MG
TML-6 is an orally active curcumin-derived research compound that inhibits synthesis of the β-amyloid precursor protein and β-amyloid (Aβ). It modulates ApoE, suppresses NF-κB and mTOR signaling, and enhances Nrf2-mediated antioxidant activity, supporting its use in Alzheimer's disease and neurodegeneration research.
  • Orally active curcumin derivative
  • Inhibits β-amyloid precursor protein and Aβ synthesis
  • Modulates ApoE expression
  • Suppresses NF-κB and mTOR signaling pathways
  • Enhances Nrf2-mediated antioxidant activity
  • Available as solid and DMSO solution pack sizes for convenience