Learn More
Medchemexpress LLC HY-137315 5mg Medchemexpress, TML-6 CAS:1462868-88-7 Purity:>98%
SDP
Catalog No. 502081484
Encompass_Preferred
Medchemexpress, HY-137315 5mg TML-6 CAS:1462868-88-7 TML-6, an orally active curcumin derivative, inhibits the synthesis of the β-amyloid precursor protein and β-amyloid (Aβ). TML-6 can upregulate Apo E, suppress NF-κB and mTOR, and increase the activity of the anti-oxidative Nrf2 gene. TML-6 has the potential for Alzheimer’s disease (AD) research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1484 Supplier Medchemexpress LLC Supplier No. HY1373155MG
Medchemexpress, HY-137315 5mg TML-6 CAS:1462868-88-7 TML-6, an orally active curcumin derivative, inhibits the synthesis of the β-amyloid precursor protein and β-amyloid (Aβ). TML-6 can upregulate Apo E, suppress NF-κB and mTOR, and increase the activity of the anti-oxidative Nrf2 gene. TML-6 has the potential for Alzheimer’s disease (AD) research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.