Learn More
Teknova TMN200 Solution, pH 8.0, Research Grade. 2L, Sterile 2 Pack.
SDP
Catalog No. 502387748
Encompass
200mM Sodium Chloride, 1mM Magnesium Chloride,20mM Tris
Catalog No. 50-238-7748 Supplier Teknova Supplier No. T14552PK
200mM Sodium Chloride, 1mM Magnesium Chloride,20mM Tris