Learn More
Aviva Systems Biology Corp TMOD2 Antibody - N-terminal region (ARP55080_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55080_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMOD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMOD2. Alternate names: MGC39481, NTMOD, N-TMODApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61418