Learn More
TARGETMOL CHEMICALS INC TMP-153 1MG
SDP
Catalog No. 502878731
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. TMP-153 is a novel and potent ACAT inhibitor with an IC50 value of 5-10 nM for intestinal and hepatic ACAT in animals.TMP-153 has a hypocholesterolemic effect, inhibiting cholesterol uptake and lowering plasma cholesterol in rats and hamsters. Purity 99.61%
Catalog No. 50-287-8731 Supplier TARGETMOL CHEMICALS INC Supplier No. T361261MG
Add to Cart
Add to Cart
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. TMP-153 is a novel and potent ACAT inhibitor with an IC50 value of 5-10 nM for intestinal and hepatic ACAT in animals.TMP-153 has a hypocholesterolemic effect, inhibiting cholesterol uptake and lowering plasma cholesterol in rats and hamsters. Purity 99.61%