Learn More
Medchemexpress LLC TMP195 | 1314891-22-9 | 99.8% | 456.42 | 250 MG
SDP
Catalog No. 5000384398
Encompass_Preferred
TMP195 is a selective class IIa histone deacetylase (HDAC) inhibitor. It has reported Ki values of 59, 60, 26, and 15 nM for HDAC4, HDAC5, HDAC7, and HDAC9, respectively. It is intended for research use only and not for sale to patients.
  • Selective class IIa HDAC inhibitor.
  • Blocks the accumulation of CCL2 protein in monocyte-derived macrophage differentiation cultures.
  • Significantly increases CCL1 protein secretion by monocytes.
  • Occupies the acetyllysine-binding site of class IIa HDACs.
  • Alters the tumor microenvironment, reducing tumor burden and pulmonary metastases by modulating macrophage phenotypes in vivo.
  • Induces recruitment and differentiation of highly phagocytic and stimulatory macrophages within tumors in vivo.
  • Combining with chemotherapy regimens or T-cell checkpoint blockade enhances the durability of tumor reduction in vivo.
Catalog No. 50-003-84398 Supplier Medchemexpress LLC Supplier No. 250MGHY18361
TMP195 is a selective class IIa histone deacetylase (HDAC) inhibitor. It has reported Ki values of 59, 60, 26, and 15 nM for HDAC4, HDAC5, HDAC7, and HDAC9, respectively. It is intended for research use only and not for sale to patients.
  • Selective class IIa HDAC inhibitor.
  • Blocks the accumulation of CCL2 protein in monocyte-derived macrophage differentiation cultures.
  • Significantly increases CCL1 protein secretion by monocytes.
  • Occupies the acetyllysine-binding site of class IIa HDACs.
  • Alters the tumor microenvironment, reducing tumor burden and pulmonary metastases by modulating macrophage phenotypes in vivo.
  • Induces recruitment and differentiation of highly phagocytic and stimulatory macrophages within tumors in vivo.
  • Combining with chemotherapy regimens or T-cell checkpoint blockade enhances the durability of tumor reduction in vivo.