Learn More
Selleck Chemical LLC TMP195 5mg 1314891-22-9 TFMO 2
SDP
Supplier:  Selleck Chemical LLC S85025MG
Encompass_Preferred
TMP195 (TFMO 2) is a selective, first-in-class, class IIa HDAC inhibitor with IC50 of 300 nM in cell-based class IIa HDAC assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8051