Learn More
Selleck Chemical LLC TMP269 10mg 1314890-29-3
SDP
Supplier:  Selleck Chemical LLC S732410MG
Encompass_Preferred
TMP269 is a potent, selective class IIa HDAC inhibitor with IC50 of 157 nM, 97 nM, 43 nM and 23 nM for HDAC4, HDAC5, HDAC7 and HDAC9, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0767607