Learn More
Medchemexpress LLC Tmp780 | 1422053-03-9 | 99.5% | 494.58 g/mol | C31H30N2O4 | 10 MG
SDP
Catalog No. 5000245660
Encompass_Preferred
TMP780 is a small-molecule inverse agonist of the retinoic acid receptor-related orphan receptor γt (RORγt), reported with an in vitro IC50 of 13 nM and used as a research tool to modulate RORγt-dependent Th17 pathways and inflammatory responses.
  • Potent RORγt inverse agonist with reported IC50 = 13 nM.
  • Useful for cellular and preclinical studies of Th17/immunoregulatory pathways.
  • Solid research compound, typically dissolved in DMSO for assays.
  • Provided in small pack sizes suitable for screening and mechanistic work.
  • CAS 1422053-03-9 enables database cross-referencing for structure and identifiers.
Catalog No. 50-002-45660 Supplier Medchemexpress LLC Supplier No. HY102075B10MG
TMP780 is a small-molecule inverse agonist of the retinoic acid receptor-related orphan receptor γt (RORγt), reported with an in vitro IC50 of 13 nM and used as a research tool to modulate RORγt-dependent Th17 pathways and inflammatory responses.
  • Potent RORγt inverse agonist with reported IC50 = 13 nM.
  • Useful for cellular and preclinical studies of Th17/immunoregulatory pathways.
  • Solid research compound, typically dissolved in DMSO for assays.
  • Provided in small pack sizes suitable for screening and mechanistic work.
  • CAS 1422053-03-9 enables database cross-referencing for structure and identifiers.