Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TMP780 | 1422053-03-9 | 99.5% | 494.58 g/mol | C31H30N2O4 | 100 MG
SDP

Catalog No. 5000245901
Encompass_Preferred

TMP780 is a small-molecule inverse agonist of the nuclear receptor RORγt used as a research compound for inflammation and immunology studies. It is reported to have potent biochemical activity and is supplied with high purity for preclinical laboratory use.

  • Inverse agonist of RORγt (IC50 13 nM).
  • High reported purity (99.51%).
  • Molecular formula C31H30N2O4; molecular weight 494.58 g/mol.
  • Typically supplied as a solid powder; soluble in DMSO.
  • Available in multiple pack sizes, including 100 mg.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-45901 Supplier Medchemexpress LLC Supplier No. HY102075B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TMP780 is a small-molecule inverse agonist of the nuclear receptor RORγt used as a research compound for inflammation and immunology studies. It is reported to have potent biochemical activity and is supplied with high purity for preclinical laboratory use.

  • Inverse agonist of RORγt (IC50 13 nM).
  • High reported purity (99.51%).
  • Molecular formula C31H30N2O4; molecular weight 494.58 g/mol.
  • Typically supplied as a solid powder; soluble in DMSO.
  • Available in multiple pack sizes, including 100 mg.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.