Learn More
Medchemexpress LLC HY-117819 5mg Medchemexpress, TMP920 CAS:1421837-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1178195MG
Encompass_Preferred
Medchemexpress, HY-117819 5mg TMP920 CAS:1421837-45-7 TMP920 is a highly potent and selective RORγt antagonist. TMP920 inhibits RORγt binding to the SRC1 peptide with an IC50 of 0.03 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9834