Learn More
Medchemexpress LLC HY-18555 5mg Medchemexpress, TMPA CAS:1258275-73-8 Purity:>98%
SDP
Catalog No. 502082045
Encompass_Preferred
Medchemexpress, HY-18555 5mg TMPA CAS:1258275-73-8 TMPA is an antagonist of nuclear receptor Nur77 and LKB1 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2045 Supplier Medchemexpress LLC Supplier No. HY185555MG
Medchemexpress, HY-18555 5mg TMPA CAS:1258275-73-8 TMPA is an antagonist of nuclear receptor Nur77 and LKB1 interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.