Learn More
Aviva Systems Biology Corp TMPO Antibody - N-terminal region (ARP44799_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44799_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPO. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMPO. Alternate names: CMD1T, LAP2, MGC61508, PRO0868, TP, LEMD4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58897