Learn More
USA Scientific Inc TempPlate semi-skirted 96-well PCR plate, straight skirt, natural, bulk
Catalog No. NC1769173
Encompass
TempPlate semi-skirted polypropylene 0.2 mL 96-well PCR plate, straight skirt, natural. 10/box.
Catalog No. NC1769173 Supplier USA Scientific Inc Supplier No. 14029220
TempPlate semi-skirted polypropylene 0.2 mL 96-well PCR plate, straight skirt, natural. 10/box.