Learn More
Electron Microscopy Sciences Tamper-Proof Cryogenic Labels Rectangle, 2.5 x 1", White, 1" core
SDP
Catalog No. 501930435
Encompass_Preferred
These labels will withstand multiple freeze-thaw cycles, boiling water and autoclaving, dry ice, UV rays, Gamma irrradiation, most chemicals and solvents, and other harsh laboratory environments.
They are ideal for prolonged storage inside liquid nitrogen tanks, freezers and dry ice as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, portable freezers and portable liquid nitrogen tanks.
Temperature range from -196 deg C to +110 deg C.
Size: 2.5 x 1"
Color: White
Core: 1"
Labels across: 1
Perforated: Yes
Catalog No. 50-193-0435 Supplier Electron Microscopy Sciences Supplier No. 7742807W
These labels will withstand multiple freeze-thaw cycles, boiling water and autoclaving, dry ice, UV rays, Gamma irrradiation, most chemicals and solvents, and other harsh laboratory environments.
They are ideal for prolonged storage inside liquid nitrogen tanks, freezers and dry ice as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, portable freezers and portable liquid nitrogen tanks.
Temperature range from -196 deg C to +110 deg C.
Size: 2.5 x 1"
Color: White
Core: 1"
Labels across: 1
Perforated: Yes