Learn More
Aviva Systems Biology Corp TMPRSS11D Antibody - N-terminal region (ARP46418_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46418_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPRSS11D. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TMPRSS11D. Alternate names: HAT, MGC150587, MGC150588ApplicationIHC, WB. Species reactivity: Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-53477