Learn More
Aviva Systems Biology Corp TMPRSS11D Antibody - middle region (ARP46419_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46419_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPRSS11D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMPRSS11D. Alternate names: HAT, MGC150587, MGC150588ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59323