Learn More
Aviva Systems Biology Corp TMPRSS12 Antibody - middle region (ARP55819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPRSS12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TMPRSS12. Alternate names: MGC57341ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-61625