Learn More
Montana Molecular TMPRSS2 BacMam - 30mL
SDP
Catalog No. NC2600307
Encompass_Preferred
TMPRSS2 packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures including A549s and HUVECs. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. This kit includes the TMPRSS2 BacMam and 500mM sodium butyrate in H2O. Use with other Montana Molecular tools including Pseudo-SARS-CoV-2 and BacMam-packaged ACE2, Neuropilin-1, and Cathepsin-L.
Catalog No. NC2600307 Supplier Montana Molecular Supplier No. C1130N030
TMPRSS2 packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures including A549s and HUVECs. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. This kit includes the TMPRSS2 BacMam and 500mM sodium butyrate in H2O. Use with other Montana Molecular tools including Pseudo-SARS-CoV-2 and BacMam-packaged ACE2, Neuropilin-1, and Cathepsin-L.