Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci TMPRSS2 (CT) Peptide
SDP

Catalog No. 16595308
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-308 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-308 Supplier ProSci Supplier No. 9569P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

TMPRSS2 (CT) Peptide (Cat. No. 9569P) is used for blocking the activity of TMPRSS2 (CT) Antibody.

Specifications

Gene ID (Entrez) 7113
Species SARS-CoV-2
Accession Number O15393
Common Name transmembrane serine protease 2
Concentration 200 μg/mL
Content And Storage Store TMPRSS2 (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol TMPRSS2
Quantity 0.05 mg
Sequence 12 amino acids near the carboxyl terminus of human TMPRSS2.
Synonym TMPRSS2: Transmembrane protease serine 2, Serine protease 10, PRSS10, Transmembrane protease serine 2 non-catalytic chain, Transmembrane protease serine 2 catalytic chain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.