Learn More
Aviva Systems Biology Corp TMPRSS3 Antibody - N-terminal region (ARP57684_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57684_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPRSS3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TMPRSS3. Alternate names: DFNB10, DFNB8, ECHOS1, TADG12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-349