Learn More
Aviva Systems Biology Corp TMPRSS4 Antibody - middle region (ARP49463_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49463_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPRSS4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TMPRSS4. Alternate names: MT-SP2, TMPRSS3, CAPH2ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-285-425