Learn More
Aviva Systems Biology Corp TMPRSS6 Antibody - N-terminal region (ARP53188_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53188_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TMPRSS6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TMPRSS6. Alternate names: IRIDAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-60962