Learn More
Surmodics Inc TMB Super Slow One Component HRP Microwell Substrate, 1L
SDP
Catalog No. NC1940234
Encompass_Preferred
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution. Peroxidase reacts with the TMB substrate solution to produce a soluble blue reaction product that can be read at 370 nm or 620-650 nm for ELISA applications. Stop solutions can be added to prevent further color development and reading the absorbance values at the desired wavelengths.
Catalog No. NC1940234 Supplier Surmodics Inc Supplier No. TTMB100001
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution. Peroxidase reacts with the TMB substrate solution to produce a soluble blue reaction product that can be read at 370 nm or 620-650 nm for ELISA applications. Stop solutions can be added to prevent further color development and reading the absorbance values at the desired wavelengths.