Learn More
Medchemexpress LLC TMPyP4 tosylate | 36951-72-1 | 98.0% | 1363.60 | 50 MG
SDP
Catalog No. 5000345222
Encompass_Preferred
TMPyP4 tosylate (TMP 1363) is identified as a quadruplex-specific ligand. It functions by inhibiting the interaction between G-quadruplexes and IGF-1. This compound also acts as a telomerase inhibitor, which in turn inhibits the proliferation of cancer cells. Furthermore, TMPyP4 tosylate serves as a stabilizer of nucleic acid secondary structure and an acetylcholinesterase inhibitor. It has also demonstrated antiviral activity against SARS-CoV-2.
  • Quadruplex-specific ligand.
  • Inhibits the interaction between G-quadruplexes and IGF-1.
  • Telomerase inhibitor.
  • Inhibits cancer cell proliferation.
  • Stabilizes nucleic acid secondary structure.
  • Acts as an acetylcholinesterase inhibitor.
  • Possesses antiviral activity against SARS-CoV-2.
Catalog No. 50-003-45222 Supplier Medchemexpress LLC Supplier No. HY10847710MG
TMPyP4 tosylate (TMP 1363) is identified as a quadruplex-specific ligand. It functions by inhibiting the interaction between G-quadruplexes and IGF-1. This compound also acts as a telomerase inhibitor, which in turn inhibits the proliferation of cancer cells. Furthermore, TMPyP4 tosylate serves as a stabilizer of nucleic acid secondary structure and an acetylcholinesterase inhibitor. It has also demonstrated antiviral activity against SARS-CoV-2.
  • Quadruplex-specific ligand.
  • Inhibits the interaction between G-quadruplexes and IGF-1.
  • Telomerase inhibitor.
  • Inhibits cancer cell proliferation.
  • Stabilizes nucleic acid secondary structure.
  • Acts as an acetylcholinesterase inhibitor.
  • Possesses antiviral activity against SARS-CoV-2.