Learn More
Abcam TMPyP4 tosylate, Telomerase inhibitor, 50MG
Catalog No. 502707258
Encompass_Preferred
MW 1363.6 g/mol, Purity >94%. Cationic porphyrin. Telomerase inhibitor (IC₅₀ = 6.5 μM). Binds and stabilizes DNA G quadruplexes resulting in telomerase inhibition. Displays antitumor effect.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7258 Supplier Abcam Supplier No. AB12079350MG
MW 1363.6 g/mol, Purity >94%. Cationic porphyrin. Telomerase inhibitor (IC₅₀ = 6.5 μM). Binds and stabilizes DNA G quadruplexes resulting in telomerase inhibition. Displays antitumor effect.
The product is subject to the following: Abcam Restricted Use Statement