Learn More
Cayman Chemical TMPyP4 (tosylate), Purity: ≥95%, CAS: 36951-72-1, 100 mg
SDP
Catalog No. 502979963
Encompass_Preferred
A cationic porphyrin that intercalates into and stabilizes G-quadruplexes; inhibits telomerase activity (IC50 ≤ 50 µM), blocking the clonogenic growth and migration of cancer cells in culture and in vivo
Catalog No. 50-297-9963 Supplier Cayman Chemical Supplier No. 18474100 MG
A cationic porphyrin that intercalates into and stabilizes G-quadruplexes; inhibits telomerase activity (IC50 ≤ 50 µM), blocking the clonogenic growth and migration of cancer cells in culture and in vivo